Tetrabutylammoniumtetraphenylborat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL281034-5GEA 103 EUR
SIAL281034-5G
Tetrabutylammoniumtetraphenylborat, Sigma-Aldrich®
Tetrabutylammonium tetraphenylborate
Formel: C₄₀H₅₆BN
MDL: MFCD00011749
CAS-Nummer: 15522-59-5
EINECS: 239-562-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR