Tetrabutylammoniumtetraphenylborat, Supelco®

Lieferant: Merck


M-Clarity™ quality level = MQ100

86897-5GEA 152 EUR
86897-5G
Tetrabutylammoniumtetraphenylborat, Supelco®
Tetrabutylammonium tetraphenylborate
Formel: C₄₀H₅₆BN
MDL: MFCD00011749
CAS-Nummer: 15522-59-5
EINECS: 239-562-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR