(2-Methylphenyl)methansulfonsäurechlorid

Lieferant: FLUOROCHEM LTD (CHEMPUR)

FLUO020884-1GEA 374 EUR
FLUO020884-1G
(2-Methylphenyl)methansulfonsäurechlorid
(2-Methylphenyl)methanesulphonyl chloride
Formel: C₈H₉ClO₂S
MDL: MFCD02683108
CAS-Nummer: 92614-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR