3-(3-Nitrophenyl)-5-phenyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6507-5GEA 291 EUR
COBBHC-6507-5G COBBHC-6507-1G
3-(3-Nitrophenyl)-5-phenyl-1,2,4-oxadiazol
3-(3-Nitrophenyl)-5-phenyl-1,2,4-oxadiazole
Formel: C₁₄H₉N₃O₃
MDL: MFCD00426946
CAS-Nummer: 20844-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR