3-(3-Nitrophenyl)-5-propyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6537-1GEA 114 EUR
COBBHC-6537-1G COBBHC-6537-5G
3-(3-Nitrophenyl)-5-propyl-1,2,4-oxadiazol
3-(3-Nitrophenyl)-5-propyl-1,2,4-oxadiazole
Formel: C₁₁H₁₁N₃O₃
MDL: MFCD10699666
CAS-Nummer: 1033202-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR