4-Fluor-N-(4-fluorbenzyl)benzamid ≥97%

Lieferant: Thermo Scientific
Achtung

H57724.MDEA 85.2 EUR
H57724.MD H57724.03
4-Fluor-N-(4-fluorbenzyl)benzamid ≥97%
4-Fluor-N-(4-fluorbenzyl)benzamid
Formel: C₁₄H₁₁F₂NO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 512181-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR