N-Benzyl-2-brombenzamid

Lieferant: COMBI-BLOCKS INC.

COBBCS-9405-1GEA 67.5 EUR
COBBCS-9405-1G COBBCS-9405-5G
N-Benzyl-2-brombenzamid
N-Benzyl-2-brombenzamid
Formel: C₁₄H₁₂BrNO
MDL: MFCD00227668
CAS-Nummer: 82082-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR