4-Benzylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS INC.

COBBAN-1507-5GEA 110 EUR
COBBAN-1507-5G COBBAN-1507-1G
4-Benzylanilin Hydrochlorid
4-Benzylanilin Hydrochlorid
Formel: C₁₃H₁₄ClN
MDL: MFCD01679497
CAS-Nummer: 6317-57-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR