3-(3-Fluor-4-methylphenyl)anilin
Lieferant: COMBI-BLOCKS INC.
COBBAN-1394-1GEA
196
EUR
COBBAN-1394-1G
COBBAN-1394-5G
3-(3-Fluor-4-methylphenyl)anilin
3-(3-Fluor-4-methylphenyl)anilin
Formel:
C₁₃H₁₂FN |
MDL:
MFCD12913928 CAS-Nummer: 1187386-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...