Tetrabutylammoniumtoluol-4-sulfonat

Lieferant: Molekula

MOLEM14693952EA 0 EUR
MOLEM14693952
Tetrabutylammoniumtoluol-4-sulfonat
Tetrabutylammoniumtoluol-4-sulfonat
Formel: C₂₃H₄₃NO₃S
Molecular Weight: 413,6574 g/mol
MDL: MFCD00043227
CAS-Nummer: 7182-86-7
EINECS: 230-548-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR