2-Methyl-2-imidazolin ≥95%, Sigma-Aldrich®

Lieferant: Merck
Achtung

SIAL375403-5GEA 136 EUR
SIAL375403-5G
2-Methyl-2-imidazolin ≥95%, Sigma-Aldrich®
2-Methyl-2-imidazolin

2-Methyl-2-imidazoline is monomethyl derivative of imidazoline. The self-association of 2-methyl-2-imidazoline was studied in aqueous solution, at different pH values, by ultraviolet spectroscopy.

Formel: C₄H₈N₂
Molecular Weight: 84,12 g/mol
MDL: MFCD00051490
CAS-Nummer: 534-26-9
EINECS: 208-596-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR