(2-Chlorphenyl)methansulfonsäurechlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL678643-1GEA 97.4 EUR
SIAL678643-1G
(2-Chlorphenyl)methansulfonsäurechlorid, Sigma-Aldrich®
(2-Chlorophenyl)methanesulphonyl chloride
Formel: C₇H₆Cl₂O₂S
MDL: MFCD04116369
CAS-Nummer: 77421-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR