3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthalinboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-2933-1GEA 490 EUR
COBBBB-2933-1G
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthalinboronsäure
3,5,5,8,8-Pentamethyl-5,6,7,8-tetrahydro-2-naphthalinboronsäure
Formel: C₁₅H₂₃BO₂
Molecular Weight: 246,1529 g/mol
MDL: MFCD08436043
CAS-Nummer: 169126-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR