1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

SIAL298743-10GEA 40.1 EUR
SIAL298743-10G
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan, Sigma-Aldrich®
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan
Formel: C₁₀H₁₆O₄
Molecular Weight: 200,23 g/mol
MDL: MFCD00010851
CAS-Nummer: 183-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR