2,8-Dimethylimidazo[1,2-a]pyridin-3-carbonsäure 95%

Lieferant: Thermo Scientific
Achtung

H50233.MDEA 57.1 EUR
H50233.MD H50233.03
2,8-Dimethylimidazo[1,2-a]pyridin-3-carbonsäure 95%
2,8-Dimethylimidazo[1,2-a]pyridin-3-carbonsäure
Formel: C₁₀H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00269632
CAS-Nummer: 874605-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR