6-Chlorimidazo[1,2-b]pyridazin Hydrochlorid

Lieferant: COMBI-BLOCKS INC.

COBBHC-6003-5GEA 34 EUR
COBBHC-6003-5G
6-Chlorimidazo[1,2-b]pyridazin Hydrochlorid
6-Chlorimidazo[1,2-b]pyridazin Hydrochlorid
Formel: C₆H₅Cl₂N₃
Molecular Weight: 190,0300 g/mol
MDL: MFCD09258637
CAS-Nummer: 13493-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR