Benzyl-(R)-(-)-glycidylether, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL363529-1GEA
35.2
EUR
SIAL363529-1G
Benzyl-(R)-(-)-glycidylether, Sigma-Aldrich®
Benzyl-(R)-(-)-glycidylether
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,20 g/mol |
MDL:
MFCD00068409 CAS-Nummer: 14618-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...