1,3-Dimethyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-8819-1GEA 60.3 EUR
COBBPN-8819-1G COBBPN-8819-250M
1,3-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
1,3-Dimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₁H₁₉BN₂O₂
MDL: MFCD09864190
CAS-Nummer: 1046832-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR