Waste containers
Lieferant: Deltalab
119-1542EA 0 EUR
DELT270045
Waste containers
Waste containers made from cardboard, reinforced, double thickness; LDPE bag 54 μm adhered to the base.
  • Safe closure with the help of raised edges
  • Printed with the biohazard anagram and text; assembling and closing instructions and drawings; identification formulary, and standards
  • Containers are supplied folded up for space-saving
Not suitable for needle disposal.
Standards: In accordance with UN ADR (road transport), manufactured pursuant to ISO 9001:2000 standards.