2-Iod-6-methoxybenzaldehyd

Lieferant: CHEMPUR

CHMP10-0001.1EA 0 EUR
CHMP10-0001.1 CHMP10-0001.500
2-Iod-6-methoxybenzaldehyd
2-Iodo-6-methoxybenzaldehyde
Formel: C₈H₇IO₂
MDL: MFCD04114363
CAS-Nummer: 5025-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR