Tetrakis(dimethylamino)zirkonium(IV), Sigma-Aldrich®

Lieferant: Merck

SIAL725528-10GEA 1210 EUR
SIAL725528-10G
Tetrakis(dimethylamino)zirkonium(IV), Sigma-Aldrich®
Tetrakis(dimethylamino)zirkonium(IV)
Formel: C₈H₂₄N₄Zr
Molecular Weight: 267,527 g/mol
MDL: MFCD00239502
CAS-Nummer: 19756-04-8
UN: 3396
ADR: 4.3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR