Tetrakis(diethylamino)zirkonium(IV), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL453153-5MLEA 292 EUR
SIAL453153-5ML
Tetrakis(diethylamino)zirkonium(IV), Sigma-Aldrich®
Tetrakis(diethylamino)zirkonium(IV)
Formel: C₁₆H₄₀N₄Zr
Molecular Weight: 379,74 g/mol
MDL: MFCD00015649
CAS-Nummer: 13801-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR