(3-Benzyloxypropyl)triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck

389196-5G
SIAL389196-5GEA 159 EUR
SIAL389196-5G
(3-Benzyloxypropyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(3-Benzyloxypropyl)triphenylphosphoniumbromid
Formel: C₂₈H₂₈BrOP
Molecular Weight: 491,41 g/mol
MDL: MFCD00191781
CAS-Nummer: 54314-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR