3-Hydroxy-2-iod-4-methoxybenzaldehyd, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL680907-5GEA
119
EUR
SIAL680907-5G
3-Hydroxy-2-iod-4-methoxybenzaldehyd, Sigma-Aldrich®
3-Hydroxy-2-iodo-4-methoxybenzaldehyde
Formel:
C₇H₈IO₃ |
MDL:
MFCD04129042 CAS-Nummer: 138490-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...