5-Methoxy-3-phenylbenzoesäure

Lieferant: COMBI-BLOCKS INC.

COBBYA-8660-1GEA 196 EUR
COBBYA-8660-1G
5-Methoxy-3-phenylbenzoesäure
5-Methoxy-3-phenylbenzoesäure
Formel: C₁₄H₁₂O₃
MDL: MFCD14702065
CAS-Nummer: 1214369-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR