Diethylphenylphosphin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL381241-1GEA
46.1
EUR
SIAL381241-1G
Diethylphenylphosphin, Sigma-Aldrich®
Diethylphenylphosphin
Formel:
C₁₀H₁₅P Molecular Weight: 166,21 g/mol |
MDL:
MFCD00015172 CAS-Nummer: 1605-53-4 EINECS: 216-516-6 UN: 3278 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...