Linalylacetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ300

SIALW263605-1KG-KEA 105 EUR
SIALW263605-1KG-K
Linalylacetat, Sigma-Aldrich®
Linalylacetat
Formel: C₁₂H₂₀O₂
MDL: MFCD00008907
CAS-Nummer: 115-95-7
EINECS: 204-116-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR