Benzyldimethyltetradecylammoniumchlorid

Lieferant: LGC Standards PROMOCHEM

PROOMM0224.02EA 1067 EUR
PROOMM0224.02
Benzyldimethyltetradecylammoniumchlorid
Benzyldimethyltetradecylammoniumchlorid
Formel: C₂₃H₄₂ClN
MDL: MFCD00011771
CAS-Nummer: 139-08-2
EINECS: 205-352-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR