(±)-2-Aminooctansäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL217700-5GEA 71.4 EUR
SIAL217700-5G
(±)-2-Aminooctansäure, Sigma-Aldrich®
(±)-2-Aminooctansäure
Formel: C₈H₁₇NO₂
Molecular Weight: 159,2261 g/mol
MDL: MFCD00008102
CAS-Nummer: 644-90-6
EINECS: 211-424-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR