Tetrakis(trimethylsiloxy)silan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL454516-5MLEA 78.1 EUR
SIAL454516-5ML
Tetrakis(trimethylsiloxy)silan, Sigma-Aldrich®
Tetrakis(trimethylsiloxy)silan
Formel: C₁₂H₃₆O₄Si₅
Molecular Weight: 384,85 g/mol
MDL: MFCD00051587
CAS-Nummer: 3555-47-3
EINECS: 222-613-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR