2-(Cyanmethoxy)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-8681-1GEA 529 EUR
COBBPN-8681-1G
2-(Cyanmethoxy)phenylboronsäurepinakolester
2-(Cyanmethoxy)phenylboronsäurepinakolester
Formel: C₁₄H₁₈BNO₃
MDL: MFCD06795686
CAS-Nummer: 936250-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR