3-Cyan-4-fluorbenzolsulfonylchlorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L19885.06EA
342
EUR
L19885.06
3-Cyan-4-fluorbenzolsulfonylchlorid ≥98%
3-Cyan-4-fluorbenzolsulfonylchlorid
Formel:
C₇H₃ClFNO₂S Molecular Weight: 219,62 g/mol Schmelzpunkt: 67…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093760 CAS-Nummer: 351003-23-1 EINECS: 000-000-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...