3-Fluor-4-hydroxy-5-nitrobenzaldehyd 95%
|
Achtung
|
H51878.03EA
189.28
EUR
H51878.03
H51878.MD
3-Fluor-4-hydroxy-5-nitrobenzaldehyd 95%
3-Fluoro-4-hydroxy-5-nitrobenzaldehyde
Formel:
C₇H₄FNO₄ Molecular Weight: 185,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846408 CAS-Nummer: 711022-47-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...