2-Chlorv-6-nitroeratrumaldehyd

Lieferant: Thermo Scientific

MAYBBTB09917.1EA 0 EUR
MAYBBTB09917.1
2-Chlorv-6-nitroeratrumaldehyd
2-Chlorv-6-nitroeratrumaldehyd
Formel: C₉H₈ClNO₅
CAS-Nummer: 82330-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR