4-Hydroxyisophthalaldehyd

Lieferant: CHEMPUR

CHMPRH01232.10EA 0 EUR
CHMPRH01232.10 CHMPRH01232.1
4-Hydroxyisophthalaldehyd
4-Hydroxyisophthalaldehyd
Formel: C₈H₆O₃
MDL: MFCD00003334
CAS-Nummer: 3328-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR