3-Amino-5-(4-methoxyphenyl)pyrazole ≥97%, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ quality level = MQ100

SIAL535370-1GEA 33 EUR
SIAL535370-1G
3-Amino-5-(4-methoxyphenyl)pyrazole ≥97%, Sigma-Aldrich®
3-Amino-5-(4-methoxyphenyl)pyrazole
Formel: C₁₀H₁₁N₃O
Molecular Weight: 189,22 g/mol
MDL: MFCD03093928
CAS-Nummer: 19541-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR