3-(4-Fluorphenyl)-1H-pyrazol-5-amin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL646725-1GEA 75.4 EUR
SIAL646725-1G SIAL646725-5G
3-(4-Fluorphenyl)-1H-pyrazol-5-amin, Sigma-Aldrich®
3-(4-Fluorphenyl)-1H-pyrazol-5-amin
Formel: C₉H₈FN₃
Molecular Weight: 177,18
MDL: MFCD01023677
CAS-Nummer: 72411-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR