D(+)-Altrose, weißes Pulver

Lieferant: MP Biomedicals

ICNA0219120810EA 36.2 EUR
ICNA0219120810
D(+)-Altrose, weißes Pulver
D(+)-Altrose

D-(+)-Altrose is used as a substrate to identify, differentiate and characterise aldose isomerases such as L-fucose isomerase from Caldicellulosiruptor saccharolyticus and d-Arabinose isomerase (d-AI) from Bacillus pallidus (B. pallidus) and Klebsiella pneumoniae.


Soluble in water (50 mg/ml - clear, colourless solution); practically insoluble in alcohol.


Bitte beachten Research Use Only (RUO).

Formel: C₆H₁₂O₆
MDL: MFCD00075620
CAS-Nummer: 1990-29-0
EINECS: 217-870-4

Bestellen

Spezifikation Testergebnisse

Identity Test Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR