1-[(Ammoniooxy)methyl]-2-nitrobenzolchlorid
Lieferant: BIONET RESEARCH (CHEMPUR)
BIOT1G-928.5EA
0
EUR
BIOT1G-928.5
BIOT1G-928.10
BIOT1G-928.500
BIOT1G-928.1
1-[(Ammoniooxy)methyl]-2-nitrobenzolchlorid
1-[(Ammoniooxy)methyl]-2-nitrobenzolchlorid
Formel:
C₇H₉ClN₂O₃ |
MDL:
MFCD01114577 CAS-Nummer: 51572-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...