1H-Benzotriazol-1-methanol

Lieferant: ASTATECH, INC.

ASTA53672EA 0 EUR
ASTA53672
1H-Benzotriazol-1-methanol
1H-Benzotriazol-1-methanol
Formel: C₇H₇N₃O
Molecular Weight: 149,1500 g/mol
MDL: MFCD00179118
CAS-Nummer: 28539-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR