1-(Trimethylsilyl)benzotriazol

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS19074.5EA 0 EUR
SILNSILS19074.5 SILNSILS19074.25 SILNSILS19074.1
1-(Trimethylsilyl)benzotriazol
1-(Trimethylsilyl)benzotriazole
Formel: C₉H₁₃N₃Si
MDL: MFCD00216577
CAS-Nummer: 43183-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR