(±)-3-Acetamidopyrrolidin

Lieferant: COMBI-BLOCKS INC.

COBBAM-1767-1GEA 74.2 EUR
COBBAM-1767-1G COBBAM-1767-5G
(±)-3-Acetamidopyrrolidin
(±)-3-Acetamidopyrrolidin
Formel: C₆H₁₂N₂O
MDL: MFCD00059019
CAS-Nummer: 79286-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR