HEPPSO Pufferlösung pH 7 (0,2 mol/l)

Lieferant: Thermo Fisher Scientific
J63877.AKEA 67.18 EUR
J63877.AK J63877.AE
HEPPSO Pufferlösung pH 7 (0,2 mol/l)
Puffer Biologische Puffer
CAS No.: 68399-78-0
Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
MDL No.: MFCD00038351
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR