Ginkgolid A
Lieferant: Thermo Scientific
J62384.FXEA
154
EUR
J62384.FX
Ginkgolid A
Ginkgolide A
Formel:
C₂₀H₂₄O₉ Molecular Weight: 408,41 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00133365 CAS-Nummer: 15291-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...