(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)

Lieferant: Apollo Scientific

APOSPC1208-100GEA 46.4 EUR
APOSPC1208-100G
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)
Formel: C₁₈H₂₈F₆N₆OP₂
Molecular Weight: 520,4 g/mol
MDL: MFCD00077411
CAS-Nummer: 128625-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR