2-Chlor-5-(trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-8242-1GEA 24.8 EUR
COBBBB-8242-1G
2-Chlor-5-(trifluormethoxy)phenylboronsäure
2-Chlor-5-(trifluormethoxy)phenylboronsäure
Formel: C₇H₅BClF₃O₃
Molecular Weight: 240,37 g/mol
MDL: MFCD06656267
CAS-Nummer: 1022922-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR