3-(4-Methylphenyl)-1H-pyrazol-5-amin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL549207-5GEA 198 EUR
SIAL549207-5G
3-(4-Methylphenyl)-1H-pyrazol-5-amin, Sigma-Aldrich®
3-(4-Methylphenyl)-1H-pyrazol-5-amin
Formel: C₁₀H₁₁N₃
MDL: MFCD00084887
CAS-Nummer: 78597-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR