Dodecacarbonyltriosmium, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL249742-1GEA 691 EUR
SIAL249742-1G
Dodecacarbonyltriosmium, Sigma-Aldrich®
Dodecacarbonyltriosmium
Formel: C₁₂O₁₂Os₃
Molecular Weight: 906,811 g/mol
MDL: MFCD00011149
CAS-Nummer: 15696-40-9
EINECS: 239-789-3
UN: 3288
ADR: 6.1,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR