Dodecacarbonyltetrarhodium, Sigma-Aldrich®
M-Clarity™ quality level = MQ100
SIAL381012-250MGEA
492
EUR
SIAL381012-250MG
Dodecacarbonyltetrarhodium, Sigma-Aldrich®
Dodecacarbonyltetrarhodium
Formel:
C₁₂O₁₂Rh₄ Molecular Weight: 747,743 g/mol |
MDL:
MFCD00016310 CAS-Nummer: 19584-30-6 EINECS: 243-171-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...