CO₂ incubator, Midi 40
Lieferant: Thermo Scientific
390-0746EA 5850 EUR
REVC3404
CO₂ incubator, Midi 40
The Midi 40 CO₂ incubator is designed specifically as a compact culturing workspace to handle small workloads, perform personal use applications, or have limited workspace. The unit has the same performance and capabilities as full-sized incubators and is a good solution to avoid shared-use environments that can present potential contamination risk through unnecessary sample handling and repeated door openings.
  • Cost-effective dedicated incubator space which prevents cross contamination
  • Removable water reservoir provides up to 95% RH with short recovery times that promote optimal cell growth
  • Even heat distribution to all sides of the chamber for highly efficient heating control
  • Standard RS485 signal output
The design features a heated inner glass door which prevents condensation, a non-porous silicon gasket maintains the chamber integrity. Units have a weldless stainless steel chamber for easy cleaning. This unit can be stacked because of its reinforced cabinet. The advanced IntrLogic™ II microprocessor has a bright digital display and intuitive touch pad for simplified entry of temperature, CO₂ and alarm data, with keyed set point switch to prevent unauthorised changes to the operating parameters. Unit has audible and visual alarm functions, precise and reliable CO₂ control featuring an accurate thermal conductivity sensor.
Lieferumfang: Supplied with four perforated stainless steel shelves.